Anti-TLR3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
042877-HRP
USBI042877-HRPEA 884 EUR
USBI042877-HRP
Anti-TLR3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human