Anti-TNFAIP8L2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
043066-BIOTIN
USBI043066-BIOTINEA 884 EUR
USBI043066-BIOTIN
Anti-TNFAIP8L2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: TNFAIP8L2 (tumor necrosis factor, alpha-induced protein 8-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR