Anti-U2AF2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
043496-BIOTIN
USBI043496-BIOTINEA 884 EUR
USBI043496-BIOTIN
Anti-U2AF2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: U2AF2 (U2 small nuclear RNA auxiliary factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human