Anti-U2AF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
043496-FITC
USBI043496-FITCEA 884 EUR
USBI043496-FITC
Anti-U2AF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: U2AF2 (U2 small nuclear RNA auxiliary factor 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human