Anti-UHRF1BP1L Rabbit Polyclonal Antibody

Lieferant: US Biological
043590
USBI043590EA 667 EUR
USBI043590
Anti-UHRF1BP1L Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: UHRF1BP1L (UHRF1 binding protein 1-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR