Anti-UNC5C Rabbit Polyclonal Antibody

Lieferant: US Biological
043613
USBI043613EA 649 EUR
USBI043613
Anti-UNC5C Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: UNC5C (unc-5 homolog C (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR