Anti-SLC7A6 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
043960-BIOTIN
USBI043960-BIOTINEA 884 EUR
USBI043960-BIOTIN
Anti-SLC7A6 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: SLC7A6 (solute carrier family 7 (amino acid transporter light chain, y+L system), member 6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR