Anti-ADIPOQ Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
044467
USBI044467EA 430 EUR
USBI044467
Anti-ADIPOQ Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen