Anti-TNFSF13B Rat Monoclonal Antibody [clone: 2.81]

Lieferant: US Biological
044578
USBI044578EA 540 EUR
USBI044578
Anti-TNFSF13B Rat Monoclonal Antibody [clone: 2.81]
Antikörper
Type: Primary
Antigen: TNFSF13B (tumor necrosis factor (ligand) superfamily, member 13b)
Clonality: Monoclonal
Clone: 2.81
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR