Anti-BAFF-R Mouse Monoclonal Antibody ATTO 488 [clone: HuBR9.1]
Lieferant: US Biological
044580
USBI044580EA 540 EUR
USBI044580
Anti-BAFF-R Mouse Monoclonal Antibody ATTO 488 [clone: HuBR9.1]
Antikörper
Type: Primary
Antigen: BAFF-R
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen