Anti-BAFF-R Mouse Monoclonal Antibody ATTO 647N [clone: HuBR9.1]

Lieferant: US Biological
044581
USBI044581EA 540 EUR
USBI044581
Anti-BAFF-R Mouse Monoclonal Antibody ATTO 647N [clone: HuBR9.1]
Antikörper
Type: Primary
Antigen: BAFF-R
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR