Anti-BAFF-R Rat Monoclonal Antibody ATTO 647N [clone: 9B9]
Lieferant: US Biological
044582
USBI044582EA 574 EUR
USBI044582
Anti-BAFF-R Rat Monoclonal Antibody ATTO 647N [clone: 9B9]
Antikörper
Type: Primary
Antigen: BAFF-R
Clonality: Monoclonal
Clone: 9B9
Conjugation: ATTO 647N
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen