Anti-Caspase-4/11, p20 Mouse Monoclonal Antibody (Biotin) [clone: Flamy-1]

Lieferant: US Biological
044636
USBI044636EA 540 EUR
USBI044636
Anti-Caspase-4/11, p20 Mouse Monoclonal Antibody (Biotin) [clone: Flamy-1]
Antikörper
Type: Primary
Antigen: Caspase-4/11, p20
Clonality: Monoclonal
Clone: Flamy-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR