Anti-DLL1 Rabbit Polyclonal Antibody

Lieferant: US Biological
044744
USBI044744EA 439 EUR
USBI044744
Anti-DLL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DLL1 (delta-like 1 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR