Anti-DLL4 Rat Monoclonal Antibody [clone: 9A1.5]

Lieferant: US Biological
044753
USBI044753EA 571 EUR
USBI044753
Anti-DLL4 Rat Monoclonal Antibody [clone: 9A1.5]
Antikörper
Type: Primary
Antigen: DLL4 (delta-like 4 (Drosophila))
Clonality: Monoclonal
Clone: 9A1.5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR