Anti-GOLGB1 Human Monoclonal Antibody ATTO 488 [clone: TA10]

Lieferant: US Biological
044838
USBI044838EA 533 EUR
USBI044838
Anti-GOLGB1 Human Monoclonal Antibody ATTO 488 [clone: TA10]
Antikörper
Type: Primary
Antigen: GOLGB1 (golgin B1)
Clonality: Monoclonal
Clone: TA10
Conjugation: ATTO 488
Epitope:
Host: Human
Isotype: IgG2 Lambda
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR