Anti-GITRL Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
044848
USBI044848EA 430 EUR
USBI044848
Anti-GITRL Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: GITRL
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR