Anti-LAG3 Mouse Monoclonal Antibody ATTO 488 [clone: 17B4]

Lieferant: US Biological
044984
USBI044984EA 608 EUR
USBI044984
Anti-LAG3 Mouse Monoclonal Antibody ATTO 488 [clone: 17B4]
Antikörper
Type: Primary
Antigen: LAG3 (lymphocyte-activation gene 3)
Clonality: Monoclonal
Clone: 17B4
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR