Anti-Rab6-GTP Human Monoclonal Antibody ATTO 488 [clone: AA2]

Lieferant: US Biological
045190
USBI045190EA 581 EUR
USBI045190
Anti-Rab6-GTP Human Monoclonal Antibody ATTO 488 [clone: AA2]
Antikörper
Type: Primary
Antigen: Rab6-GTP
Clonality: Monoclonal
Clone: AA2
Conjugation: ATTO 488
Epitope:
Host: Human
Isotype: IgG2 Lambda
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR