Anti-TNFRSF10A Mouse Monoclonal Antibody ATTO 488 [clone: HS101]

Lieferant: US Biological
045346
USBI045346EA 608 EUR
USBI045346
Anti-TNFRSF10A Mouse Monoclonal Antibody ATTO 488 [clone: HS101]
Antikörper
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: Monoclonal
Clone: HS101
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR