Anti-TNFRSF10B Mouse Monoclonal Antibody ATTO 647N [clone: HS201]

Lieferant: US Biological
045355
USBI045355EA 594 EUR
USBI045355
Anti-TNFRSF10B Mouse Monoclonal Antibody ATTO 647N [clone: HS201]
Antikörper
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: Monoclonal
Clone: HS201
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR