Anti-TUBA1A Human Monoclonal Antibody ATTO 488 [clone: F2C]

Lieferant: US Biological
045379
USBI045379EA 506 EUR
USBI045379
Anti-TUBA1A Human Monoclonal Antibody ATTO 488 [clone: F2C]
Antikörper
Type: Primary
Antigen: TUBA1A (tubulin, alpha 1a)
Clonality: Monoclonal
Clone: F2C
Conjugation: ATTO 488
Epitope:
Host: Human
Isotype: IgG2 Lambda
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR