Anti-ABCD3 Mouse Monoclonal Antibody [clone: 2H6]

Lieferant: US Biological
122812
USBI122812EA 623 EUR
USBI122812
Anti-ABCD3 Mouse Monoclonal Antibody [clone: 2H6]
Antikörper
Type: Primary
Antigen: ABCD3 (ATP-binding cassette, sub-family D (ALD), member 3)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR