Anti-ACAD8 Mouse Polyclonal Antibody

Lieferant: US Biological
122844
USBI122844EA 623 EUR
USBI122844
Anti-ACAD8 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACAD8 (acyl-CoA dehydrogenase family, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR