Anti-AKIRIN2 Mouse Monoclonal Antibody [clone: 3D9]

Lieferant: US Biological
123150
USBI123150EA 655 EUR
USBI123150
Anti-AKIRIN2 Mouse Monoclonal Antibody [clone: 3D9]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: AKIRIN2 (akirin 2)
Clonality: monoclonal
Clone: 3D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR