Anti-AKR1C3 Mouse Polyclonal Antibody

Lieferant: US Biological
123163
USBI123163EA 623 EUR
USBI123163
Anti-AKR1C3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR