Anti-ALG12 Mouse Monoclonal Antibody [clone: 5E3]

Lieferant: US Biological
123229
USBI123229EA 623 EUR
USBI123229
Anti-ALG12 Mouse Monoclonal Antibody [clone: 5E3]
Antikörper
Type: Primary
Antigen: ALG12 (ALG12, alpha-1,6-mannosyltransferase)
Clonality: monoclonal
Clone: 5000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR