Anti-ALLC Mouse Monoclonal Antibody [clone: 3D3]

Lieferant: US Biological
123242
USBI123242EA 655 EUR
USBI123242
Anti-ALLC Mouse Monoclonal Antibody [clone: 3D3]
Antikörper
Type: Primary
Antigen: ALLC (allantoicase)
Clonality: monoclonal
Clone: 3D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR