Anti-ARMC6 Mouse Monoclonal Antibody [clone: 2H8]
Lieferant: US Biological
123547
USBI123547EA 623 EUR
USBI123547
Anti-ARMC6 Mouse Monoclonal Antibody [clone: 2H8]
Antikörper
Type: Primary
Antigen: ARMC6 (armadillo repeat containing 6)
Clonality: monoclonal
Clone: 2H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human