Anti-STS Rabbit Polyclonal Antibody

Lieferant: US Biological
123589
USBI123589EA 696 EUR
USBI123589
Anti-STS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STS (steroid sulfatase (microsomal), isozyme S)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR