Anti-ASNA1 Mouse Monoclonal Antibody [clone: 2H3]

Lieferant: US Biological
123626
USBI123626EA 623 EUR
USBI123626
Anti-ASNA1 Mouse Monoclonal Antibody [clone: 2H3]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 0.5 ?g/ml

Type: Primary
Antigen: ASNA1 (arsA arsenite transporter, ATP-binding, homolog 1 (bacterial))
Clonality: monoclonal
Clone: 2H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR