Anti-ATP6V1B2 Mouse Monoclonal Antibody [clone: M1]

Lieferant: US Biological
123726
USBI123726EA 623 EUR
USBI123726
Anti-ATP6V1B2 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: ATP6V1B2 (ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR