Anti-BAZ1A Mouse Monoclonal Antibody [clone: 1F6]

Lieferant: US Biological
123843
USBI123843EA 623 EUR
USBI123843
Anti-BAZ1A Mouse Monoclonal Antibody [clone: 1F6]
Antikörper
Type: Primary
Antigen: BAZ1A (bromodomain adjacent to zinc finger domain, 1A)
Clonality: monoclonal
Clone: 1F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR