Anti-BBS7 Mouse Monoclonal Antibody [clone: 2H6]

Lieferant: US Biological
123855
USBI123855EA 623 EUR
USBI123855
Anti-BBS7 Mouse Monoclonal Antibody [clone: 2H6]
Antikörper
Type: Primary
Antigen: BBS7 (Bardet-Biedl syndrome 7)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR