Anti-BCAT2 Mouse Polyclonal Antibody

Lieferant: US Biological
123874
USBI123874EA 623 EUR
USBI123874
Anti-BCAT2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCAT2 (branched chain amino-acid transaminase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR