Anti-BHMT Mouse Polyclonal Antibody

Lieferant: US Biological
123927
USBI123927EA 623 EUR
USBI123927
Anti-BHMT Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR