Anti-BHMT2 Mouse Monoclonal Antibody [clone: 1E8]

Lieferant: US Biological
123931
USBI123931EA 623 EUR
USBI123931
Anti-BHMT2 Mouse Monoclonal Antibody [clone: 1E8]
Antikörper
Type: Primary
Antigen: BHMT2 (betaine--homocysteine S-methyltransferase 2)
Clonality: monoclonal
Clone: 100000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR