Anti-CAMP Mouse Polyclonal Antibody

Lieferant: US Biological
124307
USBI124307EA 623 EUR
USBI124307
Anti-CAMP Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR