Anti-CEP85 Mouse Polyclonal Antibody

Lieferant: US Biological
124426
USBI124426EA 655 EUR
USBI124426
Anti-CEP85 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CEP85 (centrosomal protein 85kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR