Anti-CCL2 Rabbit Polyclonal Antibody

Lieferant: US Biological
124454
USBI124454EA 662 EUR
USBI124454
Anti-CCL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR