Anti-CD28 Mouse Polyclonal Antibody

Lieferant: US Biological
124596
USBI124596EA 655 EUR
USBI124596
Anti-CD28 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR