Anti-CDKN1B Mouse Monoclonal Antibody [clone: 4B4-E6]

Lieferant: US Biological
124808
USBI124808EA 623 EUR
USBI124808
Anti-CDKN1B Mouse Monoclonal Antibody [clone: 4B4-E6]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: monoclonal
Clone: 4B4-E6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR