Anti-CHN1 Mouse Monoclonal Antibody [clone: 3A3]

Lieferant: US Biological
124963
USBI124963EA 623 EUR
USBI124963
Anti-CHN1 Mouse Monoclonal Antibody [clone: 3A3]
Antikörper
Type: Primary
Antigen: CHN1 (chimerin 1)
Clonality: monoclonal
Clone: 3A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR