Anti-CFI Mouse Polyclonal Antibody

Lieferant: US Biological
125207
USBI125207EA 623 EUR
USBI125207
Anti-CFI Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CFI (complement factor I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR