Anti-CFI Mouse Monoclonal Antibody [clone: 1B3]

Lieferant: US Biological
125209
USBI125209EA 623 EUR
USBI125209
Anti-CFI Mouse Monoclonal Antibody [clone: 1B3]
Antikörper
Type: Primary
Antigen: CFI (complement factor I)
Clonality: monoclonal
Clone: 1B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR