Anti-CRYM Rabbit Polyclonal Antibody

Lieferant: US Biological
125376
USBI125376EA 662 EUR
USBI125376
Anti-CRYM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRYM (crystallin, mu)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR