Anti-CTPS1 Mouse Monoclonal Antibody [clone: 2G7-1D10]

Lieferant: US Biological
125460
USBI125460EA 655 EUR
USBI125460
Anti-CTPS1 Mouse Monoclonal Antibody [clone: 2G7-1D10]
Antikörper
Type: Primary
Antigen: CTPS1 (CTP synthase 1)
Clonality: monoclonal
Clone: 2G7-1D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR