Anti-CTPS2 Rabbit Polyclonal Antibody

Lieferant: US Biological
125461
USBI125461EA 696 EUR
USBI125461
Anti-CTPS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTPS2 (CTP synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR