Anti-CXCL5 Mouse Monoclonal Antibody [clone: M1]

Lieferant: US Biological
125507
USBI125507EA 623 EUR
USBI125507
Anti-CXCL5 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: CXCL5 (chemokine (C-X-C motif) ligand 5)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR