Anti-CYP27A1 Rabbit Polyclonal Antibody

Lieferant: US Biological
125569
USBI125569EA 662 EUR
USBI125569
Anti-CYP27A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP27A1 (cytochrome P450, family 27, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR